Assay Details
| Gene Symbol: Mob1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026735.2 | NP_081011.1 | 4 - 5 | 651 | 145 | |
| GenBank mRNA | AK077320.1 | - | 4 - 5 | 627 | 145 | |
| AK150016.1 | - | 4 - 5 | 644 | 145 | ||
| AK162120.1 | - | 4 - 5 | 651 | 145 | ||
| AK172291.1 | - | 4 - 5 | 619 | 145 | ||
| BC104402.1 | - | 5 - 6 | 482 | 40050502 | 145 | |
| BC104403.1 | - | 4 - 5 | 546 | 40050503 | 145 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mob1b
Genomic Map