Assay Details
| Gene Symbol: Gorasp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028976.2 | NP_083252.1 | 6 - 7 | 810 | 62 | |
| GenBank mRNA | AK154986.1 | - | 6 - 7 | 787 | 62 | |
| AK170627.1 | - | 6 - 7 | 774 | 62 | ||
| AK196847.1 | - | 1 - 2 | 151 | 62 | ||
| BC012251.1 | - | 6 - 7 | 810 | 4166619 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gorasp1
Genomic Map