Assay Details
| Gene Symbol: Oxsr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133985.2 | NP_598746.2 | 15 - 16 | 1735 | 130 | |
| NR_045693.1 | 16 - 17 | 1864 | 130 | |||
| GenBank mRNA | AK031790.1 | - | 14 - 15 | 1232 | 130 | |
| AK075837.1 | - | 14 - 15 | 1243 | 130 | ||
| AK129291.1 | - | 15 - 16 | 1730 | 130 | ||
| BC036148.1 | - | 6 - 7 | 485 | 130 | ||
| BC046453.1 | - | 16 - 17 | 1759 | 130 | ||
| BC060645.1 | - | 15 - 16 | 1718 | 6852021 | 130 | |
More Information
Updated Mapping Information
Set Membership: |
|

Oxsr1
Genomic Map