Assay Details
| Gene Symbol: Oxsr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133985.2 | NP_598746.2 | 5 - 6 | 847 | 66 | |
| NR_045693.1 | 6 - 7 | 976 | 66 | |||
| GenBank mRNA | AK031790.1 | - | 4 - 5 | 344 | 66 | |
| AK075837.1 | - | 4 - 5 | 355 | 66 | ||
| AK129291.1 | - | 5 - 6 | 841 | 67 | ||
| BC060645.1 | - | 5 - 6 | 830 | 6852021 | 66 | |
More Information
Set Membership: |
|

Oxsr1
Genomic Map