Assay Details
| Gene Symbol: Phactr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005740.1 | NP_001005740.1 | 9 - 10 | 1671 | 68 | |
| NM_001005748.2 | NP_001005748.1 | 1 - 2 | 213 | 68 | ||
| GenBank mRNA | AK048208.1 | - | 9 - 10 | 1567 | 68 | |
| AK139452.1 | - | 9 - 10 | 1643 | 68 | ||
| AK166463.1 | - | 1 - 2 | 196 | 68 | ||
| AY993932.1 | - | 10 - 11 | 1608 | 68 | ||
| BC025873.1 | - | 175 | 68 | |||
| BC043065.1 | - | 4 - 5 | 697 | 68 | ||
| BC048407.1 | - | 8 - 9 | 1502 | 68 | ||
| BC061691.1 | - | 9 - 10 | 1671 | 6307669 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Phactr1
Genomic Map