Assay Details
| Gene Symbol: Edn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010104.3 | NP_034234.1 | 3 - 4 | 1018 | 87 | |
| GenBank mRNA | AB081657.1 | - | 3 - 4 | 404 | 87 | |
| AK040778.1 | - | 3 - 4 | 676 | 87 | ||
| AK152647.1 | - | 3 - 4 | 669 | 87 | ||
| AK153437.1 | - | 3 - 4 | 673 | 87 | ||
| BC029547.1 | - | 3 - 4 | 812 | 5059522 | 87 | |
| D43775.1 | - | 3 - 4 | 729 | 87 | ||
| U35233.1 | - | 3 - 4 | 423 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Edn1
Genomic Map