| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0900013388.mm.1 |
NM_026892.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900013388.mm.1 |
NM_026892.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900013388.mm.1 |
TC0900001603.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900013389.mm.1 |
NM_026892.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900013389.mm.1 |
NM_026892.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900013389.mm.1 |
TC0900001603.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900013390.mm.1 |
NM_026892.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900013390.mm.1 |
NM_026892.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900013390.mm.1 |
TC0900001603.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900013391.mm.1 |
NM_026892.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900013391.mm.1 |
NM_026892.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900013391.mm.1 |
TC0900001603.mm.1 |
MTA TC |