Assay Details
| Gene Symbol: Tert | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009354.1 | NP_033380.1 | 10 - 11 | 2664 | 83 | |
| GenBank mRNA | AF051911.1 | - | 10 - 11 | 2664 | 83 | |
| AF073311.1 | - | 10 - 11 | 2635 | 83 | ||
| BC082327.1 | - | 11 - 12 | 2166 | 83 | ||
| BC127068.1 | - | 11 - 12 | 1952 | 40110173 | 83 | |
More Information
Set Membership: |
|

Tert
Genomic Map