Assay Details
| Gene Symbol: Tert | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009354.1 | NP_033380.1 | 12 - 13 | 2974 | 101 | |
| GenBank mRNA | AF051911.1 | - | 12 - 13 | 2974 | 101 | |
| AF073311.1 | - | 12 - 13 | 2945 | 101 | ||
| BC082327.1 | - | 13 - 14 | 2476 | 101 | ||
| BC127068.1 | - | 13 - 14 | 2262 | 40110173 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tert
Genomic Map