Assay Details
| Gene Symbol: Comp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016685.2 | NP_057894.2 | 5 - 6 | 548 | 66 | |
| GenBank mRNA | AF033530.1 | - | 5 - 6 | 538 | 66 | |
| BC128368.1 | - | 5 - 6 | 538 | 40111643 | 66 | |
| BC128369.1 | - | 5 - 6 | 537 | 66 | ||
More Information
Set Membership: |
|

Comp
Genomic Map