Assay Details
| Gene Symbol: Comp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016685.2 | NP_057894.2 | 9 - 10 | 994 | 97 | |
| GenBank mRNA | AF033530.1 | - | 9 - 10 | 984 | 97 | |
| BC128368.1 | - | 9 - 10 | 984 | 40111643 | 97 | |
| BC128369.1 | - | 9 - 10 | 983 | 97 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Comp
Genomic Map