Assay Details
| Gene Symbol: Ddx49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024922.2 | NP_001020093.2 | 4 - 5 | 460 | 65 | |
| GenBank mRNA | AK139073.1 | - | 4 - 5 | 472 | 65 | |
| AK214595.1 | - | 2 - 3 | 185 | 65 | ||
| BC085301.1 | - | 4 - 5 | 460 | 65 | ||
| BC099554.1 | - | 4 - 5 | 467 | 30308124 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ddx49
Genomic Map