Assay Details
| Gene Symbol: Ddx49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024922.2 | NP_001020093.2 | 7 - 8 | 870 | 74 | |
| GenBank mRNA | BC085301.1 | - | 7 - 8 | 870 | 74 | |
| BC099554.1 | - | 7 - 8 | 877 | 30308124 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ddx49
Genomic Map