Assay Details
| Gene Symbol: Ddx49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024922.2 | NP_001020093.2 | 9 - 10 | 1047 | 106 | |
| GenBank mRNA | BC099554.1 | - | 9 - 10 | 1054 | 30308124 | 106 |
More Information
Updated Mapping Information
Set Membership: |
|

Ddx49
Genomic Map