Assay Details
| Gene Symbol: Tmem59l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182991.2 | NP_892036.1 | 7 - 8 | 920 | 89 | |
| GenBank mRNA | AK005155.1 | - | 7 - 8 | 883 | 89 | |
| BC056172.1 | - | 7 - 8 | 920 | 5699902 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25952181 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25952186 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25952170 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25952185 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25952168 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25952166 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25952176 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25952182 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25952174 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25952178 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25952169 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25952175 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25952177 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P215038 | NM_182991.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P215038 | AK005155.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P215038 | BC056172.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P215038 | NM_182991.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P215038 | AK005155.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P215038 | BC056172.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 695199 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P178688 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P215038 | NM_182991.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P215038 | AK005155.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P215038 | BC056172.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P215038 | NM_182991.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P215038 | AK005155.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P215038 | BC056172.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25952209 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25952157 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25952192 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25952208 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25952181 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25952204 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25952198 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25952196 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25952205 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25952173 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25952154 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25952191 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25952164 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25952151 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25952180 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25952159 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25952166 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25952152 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25952150 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25952161 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25952199 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25952189 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25952155 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25952178 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25952182 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25952174 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25952170 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25952186 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25952207 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25952190 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25952160 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25952194 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25952203 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25952176 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25952187 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25952197 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25952163 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25952183 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25952162 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P178688 | - | - |
| Whole Mouse Genome G4122A | A_51_P215038 | NM_182991.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P215038 | AK005155.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P215038 | BC056172.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P215038 | NM_182991.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P215038 | AK005155.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P215038 | BC056172.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P120282 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P178688 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06970990 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06806578 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06970993 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06806587 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06970987 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06806590 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25952181 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25952170 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25952174 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25952176 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25952175 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25952168 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25952169 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25952177 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25952186 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25952178 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25952166 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25952185 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25952182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952094 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25952213 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25952180 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25952160 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25952148 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25952183 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06806569 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06689110 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25952152 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25952159 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06806577 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25952181 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06806579 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25952144 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25952187 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25952151 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25952143 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25952174 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25952157 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06957236 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25952138 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25952166 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25952164 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25952161 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25952163 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25952150 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06806578 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10579289 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10579291 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10579290 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10579288 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10579293 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10579295 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10579294 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10579292 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452825_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10579287 | - | - |
| Mouse Genome 430 2.0 Array | 1452825_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1452825_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67937_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67937_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800018135.mm.1 | NM_182991.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800018135.mm.1 | NM_182991.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800018135.mm.1 | TC0800002393.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1452825_PM_at | - | - |
| Mouse Expression Set 430 | 1452825_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1452825_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1452825_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 390693 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4540170 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4540170 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004540170 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2698911 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2698911 | - | - |
| mouse-6 v1.1 (Illumina) | scl34700.7.1_46-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4540170 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl34700.7.1_46-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4540170 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2698911 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2698911 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl34700.7.1_46-S | - | - |

Tmem59l
Genomic Map