Assay Details
| Gene Symbol: Cdcp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133974.3 | NP_598735.2 | 1 - 2 | 206 | 114 | |
| GenBank mRNA | AK035674.1 | - | 1 - 2 | 168 | 114 | |
| AK087108.1 | - | 1 - 2 | 104 | 114 | ||
| BC018410.1 | - | 1 - 2 | 217 | 114 | ||
| BC059937.1 | - | 1 - 2 | 113 | 114 | ||
| BC085253.1 | - | 1 - 2 | 206 | 5347122 | 114 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdcp1
Genomic Map