Assay Details
| Gene Symbol: Cdcp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133974.3 | NP_598735.2 | 5 - 6 | 1371 | 83 | |
| GenBank mRNA | AK087108.1 | - | 5 - 6 | 1271 | 83 | |
| BC018410.1 | - | 5 - 6 | 1382 | 83 | ||
| BC059937.1 | - | 5 - 6 | 1278 | 83 | ||
| BC085253.1 | - | 5 - 6 | 1371 | 5347122 | 83 | |
More Information
Set Membership: |
|

Cdcp1
Genomic Map