Assay Details
| Gene Symbol: Cdcp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133974.3 | NP_598735.2 | 7 - 8 | 2129 | 67 | |
| GenBank mRNA | AK087108.1 | - | 7 - 8 | 2029 | 67 | |
| BC011340.1 | - | 2 - 3 | 653 | 67 | ||
| BC018410.1 | - | 7 - 8 | 2140 | 67 | ||
| BC039753.1 | - | 1 - 2 | 434 | 67 | ||
| BC059937.1 | - | 7 - 8 | 2036 | 67 | ||
| BC085253.1 | - | 7 - 8 | 2129 | 5347122 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdcp1
Genomic Map