Assay Details
| Gene Symbol: Mtrr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK028628.1 | - | 14 - 15 | 2041 | 121 | |
| AK155359.1 | - | 15 - 16 | 2754 | 121 | ||
| AK162978.1 | - | 13 - 14 | 1660 | 121 | ||
| AK163449.1 | - | 14 - 15 | 2023 | 121 | ||
| BC025942.1 | - | 1996 | 5132457 | 121 | ||
| BC031565.1 | - | 14 - 15 | 2037 | 121 | ||
| BC066796.1 | - | 13 - 14 | 1984 | 121 | ||
More Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29110675 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29110674 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29110676 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P318371 | AK028628.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P318371 | NM_172480.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P318371 | AK028628.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P318371 | NM_172480.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 181570 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 323818 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 364368 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 611678 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 797502 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 248726 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 708423 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 387649 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 699762 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 556503 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 541251 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 234878 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 279387 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 63799 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 177223 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P136053 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P165567 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P318371 | AK028628.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P318371 | NM_172480.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P318371 | AK028628.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P318371 | NM_172480.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29110647 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29110670 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29110643 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29110642 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29110640 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29110675 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29110659 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29110654 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29110645 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29110667 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29110676 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29110655 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29110666 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29110646 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29110660 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29110661 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29110652 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29110664 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29110663 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29110649 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29110641 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29110671 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29110658 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29110650 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29110656 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P165567 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P136053 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07906630 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02711986 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02711974 | - | - |
| Whole Mouse Genome G4122A | A_52_P58318 | AK028628.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P58318 | NM_172480.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P296208 | AK163449.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P296208 | AK162978.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P296208 | AK028628.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P296208 | NM_172480.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P318371 | AK028628.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P318371 | NM_172480.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P318371 | AK028628.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P318371 | NM_172480.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139141 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P136053 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P02711986 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P165567 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10921717 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10875616 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10875613 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10875618 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10921695 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10875620 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29110675 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29110676 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29110674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110579 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29110676 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29110660 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29110658 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29110650 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29110649 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29110632 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29110638 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29110626 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29110661 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29110643 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29110636 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29110652 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10958635 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10875613 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29110646 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29110655 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29110659 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29110670 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29110621 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29110634 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10958617 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29110666 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29110639 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29110622 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29110654 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10958628 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29110663 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29110629 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29110631 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10875620 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29110633 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10958641 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10958623 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10657313 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29110671 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10958637 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29110667 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10410405 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10410399 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10410401 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10410404 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10410396 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10410400 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10410406 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10410395 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10410402 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10410394 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10410397 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10410403 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10410407 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10410398 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452110_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10410393 | - | - |
| Mouse Genome 430 2.0 Array | 1452110_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1452110_at | - | - |
| Mouse Genome 430 2.0 Array | 1458093_at | - | - |
| Mouse Genome 430 2.0 Array | 1458093_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 210009_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 210009_PM_at | - | - |
| Mouse Expression Set 430 | 1452110_PM_at | - | - |
| Mouse Expression Set 430 | 1452110_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452110_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452110_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452110_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452110_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5910373 | - | - |
| MouseWG-6 v2 Expression BeadChip | 6180397 | - | - |
| MouseWG-6 v2 Expression BeadChip | 4060358 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5910373 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6180397 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4060358 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006180397 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005910373 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004060358 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2732767 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2742268 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2723383 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2723383 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2742268 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2732767 | - | - |
| mouse-6 v1.1 (Illumina) | scl00210009.1_92-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0003635.1_71-S | - | - |
| mouse-6 v1.1 (Illumina) | scl00210009.2_325-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4060358 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5910373 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6180397 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00210009.1_92-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00210009.2_325-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0003635.1_71-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5910373 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4060358 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6180397 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2723383 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2969314 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2969317 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2742268 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2742268 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2969317 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2723383 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2969314 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0003635.1_71-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl00210009.1_92-S | - | - |

Mtrr
Genomic Map