Assay Details
| Gene Symbol: Mtrr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK028628.1 | - | 5 - 6 | 871 | 62 | |
| AK155359.1 | - | 6 - 7 | 1584 | 62 | ||
| AK163449.1 | - | 5 - 6 | 853 | 62 | ||
| BC025942.1 | - | 826 | 5132457 | 62 | ||
| BC031565.1 | - | 5 - 6 | 867 | 62 | ||
| BC066796.1 | - | 4 - 5 | 814 | 62 | ||
More Information
Set Membership: |
|

Mtrr
Genomic Map