Assay Details
| Gene Symbol: Mtrr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK028628.1 | - | 8 - 9 | 1229 | 97 | |
| AK155359.1 | - | 9 - 10 | 1942 | 97 | ||
| AK162978.1 | - | 7 - 8 | 848 | 97 | ||
| AK163449.1 | - | 8 - 9 | 1211 | 97 | ||
| BC025942.1 | - | 1184 | 5132457 | 97 | ||
| BC031565.1 | - | 8 - 9 | 1225 | 97 | ||
| BC066796.1 | - | 7 - 8 | 1172 | 97 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mtrr
Genomic Map