Assay Details
| Gene Symbol: Ebp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK012266.1 | - | 1 - 2 | 99 | 74 | |
| AK027977.1 | - | 1 - 2 | 71 | 74 | ||
| BC004620.1 | - | 1 - 2 | 77 | 3585323 | 74 | |
| BC004703.1 | - | 1 - 2 | 709 | 3499265 | 74 | |
| X97755.1 | - | 1 - 2 | 75 | 74 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK012266.1, AK027977.1, BC004620.1, BC004703.1, X97755.1 transcripts.
Set Membership: |
|

Ebp
Genomic Map