Assay Details
| Gene Symbol: Naf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163564.1 | NP_001157036.1 | 5 - 6 | 1331 | 97 | |
| GenBank mRNA | AK144735.1 | - | 5 - 6 | 578 | 97 | |
| BC027148.1 | - | 1 - 2 | 114 | 97 | ||
| BC053440.1 | - | 1 - 2 | 102 | 97 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Naf1
Genomic Map