Assay Details
| Gene Symbol: Galntl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175032.3 | NP_778197.2 | 10 - 11 | 1642 | 66 | |
| GenBank mRNA | GU220061.1 | - | 10 - 11 | 1489 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Galntl6
Genomic Map