Assay Details
| Gene Symbol: Glb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009752.2 | NP_033882.1 | 10 - 11 | 1188 | 107 | |
| NR_108102.1 | 9 - 10 | 1018 | 107 | |||
| GenBank mRNA | AK042069.1 | - | 10 - 11 | 1158 | 107 | |
| AK048460.1 | - | 10 - 11 | 1119 | 107 | ||
| AK076517.1 | - | 10 - 11 | 1159 | 107 | ||
| AK153218.1 | - | 10 - 11 | 1170 | 107 | ||
| AK171593.1 | - | 10 - 11 | 1191 | 107 | ||
| AK171760.1 | - | 9 - 10 | 989 | 107 | ||
| AK204903.1 | - | 2 - 3 | 166 | 107 | ||
| BC028875.1 | - | 10 - 11 | 1108 | 4240285 | 107 | |
| M57734.1 | - | 10 - 11 | 1147 | 107 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Glb1
Genomic Map