Assay Details
| Gene Symbol: Tgfbr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009371.3 | NP_033397.3 | 1 - 2 | 419 | 70 | |
| GenBank mRNA | AK043619.1 | - | 1 - 2 | 503 | 70 | |
| AK053793.1 | - | 1 - 2 | 420 | 70 | ||
| AK159327.1 | - | 1 - 2 | 399 | 70 | ||
| BC052629.1 | - | 321 | 70 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Tgfbr2
Genomic Map