Assay Details
| Gene Symbol: Ripk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009068.3 | NP_033094.3 | 3 - 4 | 457 | 79 | |
| GenBank mRNA | AK030959.1 | - | 3 - 4 | 688 | 79 | |
| AK048989.1 | - | 3 - 4 | 685 | 79 | ||
| AK156803.1 | - | 3 - 4 | 722 | 79 | ||
| BC050905.1 | - | 3 - 4 | 457 | 5721177 | 79 | |
| BC054542.1 | - | 3 - 4 | 457 | 5721177 | 79 | |
| BC058162.1 | - | 3 - 4 | 643 | 5135235 | 79 | |
| U25995.1 | - | 2 - 3 | 372 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ripk1
Genomic Map