Assay Details
| Gene Symbol: Padi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011059.2 | NP_035189.1 | 13 - 14 | 1672 | 98 | |
| GenBank mRNA | AB013848.1 | - | 13 - 14 | 1672 | 98 | |
| AK037182.1 | - | 13 - 14 | 1673 | 98 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Padi1
Genomic Map