Assay Details
| Gene Symbol: Padi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011059.2 | NP_035189.1 | 8 - 9 | 1050 | 133 | |
| GenBank mRNA | AK037182.1 | - | 8 - 9 | 1051 | 133 | |
More Information
Set Membership: |
|

Padi1
Genomic Map