Assay Details
| Gene Symbol: Arl13a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028947.1 | NP_083223.1 | 1 - 2 | 156 | 106 | |
| GenBank mRNA | AK016872.1 | - | 1 - 2 | 157 | 106 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK016872.1, NM_028947.1 transcripts.
Set Membership: |
|

Arl13a
Genomic Map