Assay Details
| Gene Symbol: Arl13a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028947.1 | NP_083223.1 | 8 - 9 | 1209 | 87 | |
| GenBank mRNA | AK016872.1 | - | 8 - 9 | 1210 | 87 | |
| BC100522.1 | - | 8 - 9 | 1107 | 30920038 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arl13a
Genomic Map