Assay Details
| Gene Symbol: Arl13a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028947.1 | NP_083223.1 | 9 - 10 | 1289 | 70 | |
| GenBank mRNA | AK016872.1 | - | 9 - 10 | 1290 | 70 | |
| BC100522.1 | - | 9 - 10 | 1187 | 30920038 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P195203 | BC100522.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P195203 | NM_028947.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P195203 | AK016872.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P195203 | BC100522.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P195203 | NM_028947.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P195203 | AK016872.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 376596 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 297637 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 899750 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 31348 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 302465 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 215741 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 951486 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 424286 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 414804 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 436945 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 688523 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 83550 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 13117 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 37200 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P154989 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P163016 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P195203 | BC100522.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P195203 | NM_028947.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P195203 | AK016872.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P195203 | BC100522.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P195203 | NM_028947.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P195203 | AK016872.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P163016 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P154989 | - | - |
| Whole Mouse Genome G4122A | A_51_P195203 | BC100522.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P195203 | NM_028947.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P195203 | AK016872.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P195203 | BC100522.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P195203 | NM_028947.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P195203 | AK016872.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P163016 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P154989 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32670933 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14928287 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32670884 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15007260 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14995103 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32670869 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14853832 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32670889 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32670904 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32670866 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32670908 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32670925 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32670924 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32670899 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32670831 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32670873 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32670932 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32670874 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32670897 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15051040 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15033903 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32670892 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32670881 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14891435 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32670931 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14891436 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32670878 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32670864 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32670851 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32670838 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32670909 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32670905 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32670900 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32670868 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32670895 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32670858 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32670844 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32670856 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32670863 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32670922 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15007267 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32670883 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32670876 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32670835 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15007265 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32670841 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32670828 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32670871 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32670839 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32670855 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32670867 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32670886 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32670845 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32670847 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32670860 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32670879 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32670843 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32670927 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32670853 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32670857 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32670906 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32670849 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14928286 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32670836 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10601696 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10601694 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10601693 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10601700 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10601697 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10601695 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10601698 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10601699 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10601692 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1431945_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10601691 | - | - |
| Mouse Genome 430 2.0 Array | 1431945_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1431945_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 74448_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 74448_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0X00007897.mm.1 | NM_028947.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00007897.mm.1 | NM_028947.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00007897.mm.1 | TC0X00001274.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1416710_PM_at | - | - |
| Mouse Expression Set 430 | 1416710_at | - | - |
| Mouse Expression Set 430 | 1431945_at | - | - |
| Mouse Expression Set 430 | 1436723_at | - | - |
| Mouse Expression Set 430 | 1431945_PM_at | - | - |
| Mouse Expression Set 430 | 1436723_PM_at | - | - |
| Murine Genome U74v2 Set | 165464_r_at | - | - |
| Murine Genome U74v2 Set | 165464_PM_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416710_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416710_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431945_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431945_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 645280 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1230103 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1230103 | - | - |

Arl13a
Genomic Map