Assay Details
| Gene Symbol: Trmt2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167994.1 | NP_001161466.1 | 5 - 6 | 640 | 76 | |
| NM_172540.2 | NP_766128.2 | 4 - 5 | 564 | 76 | ||
| GenBank mRNA | AK028999.1 | - | 4 - 5 | 520 | 76 | |
| AK049523.1 | - | 4 - 5 | 562 | 76 | ||
| AK129030.1 | - | 5 - 6 | 583 | 76 | ||
| AK137595.1 | - | 526 | 76 | |||
| BC030387.1 | - | 3 - 4 | 225 | 76 | ||
| BC056937.1 | - | 5 - 6 | 581 | 5716144 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trmt2b
Genomic Map