Assay Details
| Gene Symbol: Srpx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026838.4 | NP_081114.2 | 1 - 2 | 354 | 89 | |
| GenBank mRNA | BB848735.1 | - | 1 - 2 | 355 | 89 | |
| BC028307.1 | - | 1 - 2 | 83 | 3488526 | 89 | |
| CN539245.1 | - | 1 - 2 | 117 | 89 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC028307.1, NM_026838.4 transcripts.
Set Membership: |
|

Srpx2
Genomic Map