Assay Details
| Gene Symbol: Srpx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083895.3 | NP_001077364.2 | 6 - 7 | 1159 | 122 | |
| NM_026838.4 | NP_081114.2 | 7 - 8 | 1211 | 122 | ||
| GenBank mRNA | AF393640.1 | - | 5 - 6 | 729 | 122 | |
| AK136935.1 | - | 6 - 7 | 1014 | 122 | ||
| BC028307.1 | - | 7 - 8 | 940 | 3488526 | 122 | |
| BQ830905.1 | - | 3 - 4 | 370 | 122 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Srpx2
Genomic Map