Assay Details
| Gene Symbol: Muc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080457.3 | NP_536705.3 | 1 - 2 | 561 | 70 | |
| GenBank mRNA | AF441786.1 | - | 172 | 70 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Muc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080457.3 | NP_536705.3 | 1 - 2 | 561 | 70 | |
| GenBank mRNA | AF441786.1 | - | 172 | 70 | ||
Set Membership: |
|