Assay Details
| Gene Symbol: 1110038B12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015536.1 | 2 - 3 | 156 | 97 | ||
| GenBank mRNA | AK002372.1 | - | 2 - 3 | 156 | 97 | |
| BC058123.1 | - | 2 - 3 | 140 | 97 | ||
| BE133139.1 | - | 2 - 3 | 97 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

1110038B12Rik
Genomic Map