Assay Details
| Gene Symbol: 1110038B12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK003073.1 | - | 2 - 3 | 154 | 111 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
small nucleolar RNA, C/D box 52 |
Snord52 | MBII-297 | -737.0 | Sense | Chr. 17 - 34950950 - 34951008 on NCBI Build GRCm38 |
| Downstream | heat shock protein 1B | Hspa1b |
Hsp70, Hsp70-1, Hsp70.1, hsp68 |
4.3kb | Sense | Chr. 17 - 34956429 - 34959238 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

1110038B12Rik
Genomic Map