Assay Details
| Gene Symbol: 3110073H01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AI606459.1 | - | 1 | 131 | ||
| AK014252.1 | - | 1 | 1864 | 131 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | S-phase kinase-associated protein 2 (p45) | Skp2 |
4930500A04Rik, FBXL1, FWD1, p45 |
-46.6kb | Sense | Chr. 15 - 9111982 - 9155425 on NCBI Build GRCm38 |
| Downstream |
UDP glycosyltransferases 3 family, polypeptide A1 |
Ugt3a1 | AI746432 | 77.3kb | Anti-sense | Chr. 15 - 9279826 - 9315038 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

3110073H01Rik
Genomic Map