Assay Details
| Gene Symbol: 4930474G06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK076809.1 | - | 1 - 2 | 55 | 98 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cDNA sequence BC051408 | BC051408 | -145.4kb | Sense | Chr. 18 - 28711861 - 28726847 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA 4930578E11 gene | 4930578E11Rik | 516.6kb | Anti-sense | Chr. 18 - 29396716 - 29496416 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

4930474G06Rik
Genomic Map