Assay Details
| Gene Symbol: Fdft1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK076062.1 | - | 1 - 2 | 167 | 89 | |
| AK136159.1 | - | 3 - 4 | 947 | 89 | ||
| AK146766.1 | - | 1 - 2 | 175 | 89 | ||
| AK158633.1 | - | 1 - 2 | 164 | 89 | ||
| AK162294.1 | - | 1 - 2 | 128 | 89 | ||
| AK168629.1 | - | 1 - 2 | 101 | 89 | ||
| AK169317.1 | - | 1 - 2 | 180 | 89 | ||
| BC054722.1 | - | 1 - 2 | 109 | 5694816 | 89 | |
| BC138301.1 | - | 1 - 2 | 163 | 8861320 | 89 | |
| BC138302.1 | - | 1 - 2 | 164 | 8861321 | 89 | |
| D29016.1 | - | 1 - 2 | 117 | 89 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Fdft1
Genomic Map