Assay Details
| Gene Symbol: Fdft1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK076062.1 | - | 2 - 3 | 267 | 72 | |
| AK146766.1 | - | 2 - 3 | 275 | 72 | ||
| AK158633.1 | - | 2 - 3 | 264 | 72 | ||
| AK162294.1 | - | 2 - 3 | 228 | 72 | ||
| AK168629.1 | - | 2 - 3 | 201 | 72 | ||
| AK169317.1 | - | 2 - 3 | 280 | 72 | ||
| BC054722.1 | - | 2 - 3 | 209 | 5694816 | 72 | |
| BC138301.1 | - | 2 - 3 | 263 | 8861320 | 72 | |
| BC138302.1 | - | 2 - 3 | 264 | 8861321 | 72 | |
| D29016.1 | - | 2 - 3 | 217 | 72 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Fdft1
Genomic Map