Assay Details
| Gene Symbol: Gdi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008112.4 | NP_032138.3 | 6 - 7 | 943 | 121 | |
| GenBank mRNA | AK007871.1 | - | 6 - 7 | 886 | 121 | |
| AK078144.1 | - | 886 | 121 | |||
| AK078514.1 | - | 6 - 7 | 890 | 121 | ||
| AK079690.1 | - | 5 - 6 | 784 | 121 | ||
| AK083063.1 | - | 6 - 7 | 885 | 121 | ||
| AK090083.1 | - | 6 - 7 | 891 | 121 | ||
| AK137761.1 | - | 6 - 7 | 890 | 121 | ||
| AK143633.1 | - | 6 - 7 | 888 | 121 | ||
| AK143681.1 | - | 6 - 7 | 922 | 121 | ||
| AK149958.1 | - | 6 - 7 | 896 | 121 | ||
| AK150356.1 | - | 6 - 7 | 878 | 121 | ||
| AK150655.1 | - | 6 - 7 | 913 | 121 | ||
| AK152250.1 | - | 6 - 7 | 877 | 121 | ||
| AK153381.1 | - | 6 - 7 | 856 | 121 | ||
| AK159734.1 | - | 6 - 7 | 888 | 121 | ||
| AK166317.1 | - | 6 - 7 | 888 | 121 | ||
| AK167659.1 | - | 6 - 7 | 900 | 121 | ||
| AK169299.1 | - | 6 - 7 | 854 | 121 | ||
| BC024971.1 | - | 6 - 7 | 864 | 121 | ||
| BC053381.1 | - | 3043 | 121 | |||
| BC055341.1 | - | 6 - 7 | 869 | 6808475 | 121 | |
| L36314.1 | - | 6 - 7 | 749 | 121 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gdi2
Genomic Map