Assay Details
| Gene Symbol: Tk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271729.1 | NP_001258658.1 | 1 - 2 | 303 | 99 | |
| GenBank mRNA | CF732600.1 | - | 1 - 2 | 118 | 99 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the NM_001271729.1 transcripts.
This assay is in the 5'UTR region of the NM_001271729.1 transcripts.
Set Membership: |
|

Tk1
Genomic Map