Assay Details
| Gene Symbol: Otud6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152812.3 | NP_690025.2 | 3 - 4 | 530 | 156 | |
| GenBank mRNA | AK144761.1 | - | 3 - 4 | 353 | 152 | |
| AK155761.1 | - | 3 - 4 | 381 | 156 | ||
| AK166445.1 | - | 3 - 4 | 360 | 156 | ||
| AK169134.1 | - | 3 - 4 | 345 | 156 | ||
| AK172229.1 | - | 3 - 4 | 335 | 156 | ||
| AK172338.1 | - | 3 - 4 | 335 | 156 | ||
| BC031474.1 | - | 3 - 4 | 336 | 2599116 | 156 | |
| BC087552.1 | - | 3 - 4 | 327 | 6331952 | 156 | |
More Information
Updated Mapping Information
Set Membership: |
|

Otud6b
Genomic Map