Assay Details
| Gene Symbol: Rpl19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009078.2 | NP_033104.2 | 1 - 2 | 67 | 75 | |
| GenBank mRNA | AK010440.1 | - | 1 - 2 | 34 | 75 | |
| AK075817.1 | - | 1 - 2 | 34 | 75 | ||
| AK168232.1 | - | 1 - 2 | 38 | 75 | ||
| AK168481.1 | - | 1 - 2 | 38 | 75 | ||
| BC087961.1 | - | 1 - 2 | 33 | 6761831 | 75 | |
| BC089549.1 | - | 1 - 2 | 34 | 6703351 | 75 | |
| BU524941.1 | - | 1 - 2 | 69 | 75 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK010440.1,AK168481.1,BC087961.1,BC089549.1,NM_009078.2 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rpl19
Genomic Map