Assay Details
| Gene Symbol: Rpl19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159483.1 | NP_001152955.1 | 3 - 4 | 285 | 59 | |
| NM_009078.2 | NP_033104.2 | 3 - 4 | 299 | 59 | ||
| GenBank mRNA | AK010440.1 | - | 3 - 4 | 266 | 59 | |
| AK075817.1 | - | 3 - 4 | 266 | 59 | ||
| AK168232.1 | - | 270 | 59 | |||
| AK168481.1 | - | 3 - 4 | 270 | 59 | ||
| BC010710.1 | - | 2 - 3 | 244 | 2648593 | 59 | |
| BC083131.1 | - | 2 - 3 | 246 | 6442192 | 59 | |
| BC087961.1 | - | 3 - 4 | 265 | 6761831 | 59 | |
| BC089549.1 | - | 3 - 4 | 266 | 6703351 | 59 | |
| BC092539.1 | - | 3 - 4 | 255 | 6594084 | 59 | |
| BU524941.1 | - | 3 - 4 | 301 | 59 | ||
| CB203522.1 | - | 3 - 4 | 285 | 59 | ||
| M62952.1 | - | 2 - 3 | 237 | 59 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rpl19
Genomic Map