Assay Details
| Gene Symbol: Ost4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024460.4 | NP_077780.3 | 1 - 2 | 118 | 89 | |
| GenBank mRNA | AA563474.1 | - | 1 - 2 | 118 | 89 | |
| AK002224.2 | - | 1 - 2 | 93 | 89 | ||
| AK003569.1 | - | 1 - 2 | 97 | 89 | ||
| AK009384.1 | - | 53 | 89 | |||
| AK019285.1 | - | 1 - 2 | 78 | 89 | ||
| CA466367.1 | - | 1 - 2 | 76 | 89 | ||
| DV041491.1 | - | 159 | 89 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the AK002224.2,AK003569.1,AK019285.1,NM_024460.4 transcripts.
This assay is in the 5'UTR region of the AK002224.2,AK003569.1,AK019285.1,NM_024460.4 transcripts.
Set Membership: |
|

Ost4
Genomic Map