Assay Details
| Gene Symbol: Calcoco2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271018.1 | NP_001257947.1 | 2 - 3 | 198 | 78 | |
| GenBank mRNA | AF490344.1 | - | 2 - 3 | 196 | 78 | |
| AK010816.1 | - | 2 - 3 | 196 | 78 | ||
| AK145542.1 | - | 2 - 3 | 199 | 78 | ||
| AK166710.1 | - | 2 - 3 | 199 | 78 | ||
| AK166756.1 | - | 2 - 3 | 199 | 78 | ||
| AK166939.1 | - | 2 - 3 | 196 | 78 | ||
| AK167009.1 | - | 2 - 3 | 199 | 78 | ||
| AK167172.1 | - | 2 - 3 | 199 | 78 | ||
| AK167291.1 | - | 2 - 3 | 198 | 78 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Calcoco2
Genomic Map