Assay Details
| Gene Symbol: 1110038B12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027943.1 | 2 - 3 | 153 | 102 | ||
| GenBank mRNA | AK004150.1 | - | 2 - 3 | 154 | 102 | |
More Information
Additional Information:
This assay targets the same location as MTA transcript cluster(s) TC1700001933.mm
This assay targets the same location as RTA transcript cluster(s) TC1000000405.rn
This assay targets the same location as RTA transcript cluster(s) TC1000000405.rn
Set Membership: |
|

1110038B12Rik
Genomic Map